Nicotine

Supplier: ALDON CORP SE

NN0195-100ML NN0195-100MLR
470301-866EA 16.9 USD
470301-866 470301-868
Nicotine
Chemicals for Science Education

Formula Weight: Mixture
Formula: Mixture
Density (g/mL): Approximately 1
Boiling Point (°C): Approximately 100
Freezing Point (°C): Approximately 0
Solubility: Water and Organic Solvents
Synonyms: Cigarette Solution
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR