Ward's® Live Mycobacterium nonchromogenicum Culture
Supplier: VWR
WARD470179-200
470179-200PK 12.95 USD
470179-200
Ward's® Live Mycobacterium nonchromogenicum Culture
Specimens Live Specimens Bacteria - Living Material
This organism is normally isolated from soil and is a non-pathogenic strain of the mycobacterium genus.
  • Biosafety Level: 1
  • Motility: Negative
  • Morphology: Slender rods
  • Gram Reaction: Positive (but not readily stained)
  • Acid Fast: Yes
  • Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: While this organism is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.