Milled Sphagnum Moss
Supplier: Avantor
WARD470014-980
470014-980PK 16.5 USD
470014-980
Milled Sphagnum Moss
Gardening Supplies
Naturally sterile, with superior water retention, the medium eliminates “damping off” for a higher seed germination rate. Excellent for starting seeds indoors. Quantity: 8 oz.