Media Bottles
Supplier: DWK Life Sciences (KIMBLE)
219815 219817 219820
470205-570CS 509.65 USD
470205-570 470205-572 470205-576
Media Bottles
Bottles Glass Bottles
Media bottles are manufactured from borosilicate glass for excellent chemical resistance. A no-drop pour lip facilicates pouring and cleaning. Graduations show approximate volume, while a large writing patch can be used for identifying content.
Each bottle has a black phenolic PTFE rubber-lined cap attached. Caps are autoclavable.