Magnetic Fingerprint Kit
1-0123
470024-158EA 96.5 USD
470024-158
Magnetic Fingerprint Kit
Educational Materials Forensics Educational Materials Fingerprint Analysis Learning Activities
Includes 90 cc Wide-Mouth Powder Jar for Convenient Use With the Magnetic Applicator.
Professional fingerprinting techniques are easy to complete and duplicate, with this comprehensive kit. Uncover latent fingerprints using a magnetic powder applicator, 1 oz. of bi–chromatic magnetic powder, one 1 1/2" x 360" roll of lifting tape, and one hundred 3" x 5" cards, all contained in a sturdy carrying case. Size: 10 3/8"L x 4 5/8"W x 1 3/4"H.
Available to U.S. customers only; similar product will ship elsewhere.