Lycopodium

Supplier: ALDON CORP SE

LL0270-25G LL0270-PK/4 LL0270-500G
470301-612EA 18.65 USD
470301-612 470301-614 470301-616
Lycopodium
Chemicals for Science Education

CAS Number: 8023-70-9
Formula: Natural Product
Solubility: Insoluble in Water and Organic Solvents
Synonyms: Spores of Lycopodium Clavatum
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is flammable.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR