Liquid Crystal Thermometer Strip
Supplier: AMERICAN EDUCATIONAL PROD
6083
470007-494EA 22.4 USD
470007-494
Liquid Crystal Thermometer Strip
Temperature Indicating Strips
Students make a temperature map of the human body with the Liquid Crystal Thermometer Strip.
  • 1 °C increments
This flexible strip is used to measure the differences in temperature across various surfaces of the skin.
Caution: Do not submerse in water.