LaMotte® Soil Micronutrients Outfit
Supplier: LA MOTTE CHEMICAL PROD CO SE
5938-02 R-5938-01
470002-174KT 818.9 USD
470002-174 470002-176
LaMotte® Soil Micronutrients Outfit
Standards Environmental Analysis Standards
Tests for Nine Essential Minor Elements
  • Students can work cooperatively
  • Tests are packaged individually
  • Test soil nutrients and metals
You’ll get enough materials for 50 tests each of aluminum, ammonia nitrogen, calcium, chlorides, iron, magnesium, manganese, nitrite nitrogen, and sulfates packed in independent modules so several teams can work simultaneously. Size: 15"L x 13"W x 7"H; weight: 19 lbs. Note: Contains barium chloride.