Kidney, Mammal Slide
Supplier: Avantor
WARD470182-924
470182-924EA 13.5 USD
470182-924
Kidney, Mammal Slide
Slides Prepared Slides
Investigate kidney structure
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. For detailed study of cortex and medulla. Shows renal corpuscle, tubule, and collecting duct.