Kalanchoe blossfeldiana
Supplier: CHASE GREENHOUSES, INC.
866910
470180-016PK 84.85 USD
470180-016
Kalanchoe blossfeldiana
Specimens Live Specimens Plants - Living Material
Dicot. Easy to grow and resistant to secondary infections, with fleshy leaves and bright flowers, this plant is often utilized as a host for plant tissue cancer studies. It also illustrates vegetative reproduction as they grow from adventitious buds. It is native to the tropics and is supplied in a pot with care instructions. Size: 4–6". Package of eight.