Iodine

Supplier: ALDON CORP SE

IX0120-PK/4 IX0120-100G IX0120-25G IX0120-500G IX0120-25GR IX0120-100GR
470301-338PK 88.65 USD
470301-338 470301-340 470301-342 470301-344 470301-348 470301-508
Iodine
Chemicals for Science Education

CAS Number: 7553-56-2
Formula Weight: 253.80
Formula: I2
Hazard Info: Corrosive, Irritant, Toxic
Density (g/mL): 4.93
Boiling Point (°C): 184
Freezing Point (°C): 113.5
Solubility: Common Organic Solvents
Shelf Life (months): 12
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR