Introductory Rock-Forming Mineral Collection
Supplier: Avantor
WARD470025-152
470025-152EA 54.59 USD
470025-152
Introductory Rock-Forming Mineral Collection
Specimens Mineral Specimens Mineral Collections
Examples of the More Important Minerals Found in Rocks
This basic set of minerals demonstrates those that can be found in igneous, sedimentary, and metamorphic rocks. It is an ideal size for beginners’ individual study and includes a study guide.Contents: 24 numbered samples, 1×1"Minerals:augite, biotite, calcite, chlorite, dolomite, epidote, garnet, gypsum, hematite, hornblende, labradorite, limonite, magnetite, microcline, muscovite, nepheline, olivine, phlogopite, pyrite, pyrrhotite, quartz (flint), quartz (rock crystal), serpentine, talc.
Caution: Due to the nature of this product it may contain asbestos.