Ward's® Introduction to Cloning Lab Activity: Duckweed
Supplier: Avantor
470030-356KT 106.49 USD
470030-356
Ward's® Introduction to Cloning Lab Activity: Duckweed
Cell Culture Media
Simple Method for Replicating Easily Cloned Plant
  • Clone an organism in your class
  • Develop aseptic technique
  • Materials for 10 lab groups
  • Time Required: 60 minutes plus ~2 weeks
Provide with a solid foundation in plant tissue culture with our easy to use kit; sterile media isn’t required, making cloning the supplied duckweed even more convenient. The activity contains enough materials for ten setups, as well as teacher’s guide and student copymaster.
No. of Lab Groups: 10
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.