Heavy Duty Beaker Tongs
Supplier: HUMBOLDT MFG CO
H-23440-LBL
470123-720EA 59.62 USD
470123-720
Heavy Duty Beaker Tongs
Tongs
Permits handling of hot beakers from 100 to 1500 mL capacity.
Stainless steel with riveted joint and fiberglass-covered jaws. Length: 12-1/2".