Antibiotic Test Disc Sets
Supplier: Avantor
231039
470178-404KT 131.95 USD
470178-404 470178-406 470178-408
Antibiotic Test Disc Sets
Media for Microbiology Prepared Media for Microbiology
Easily observe the sensitivity of various bacteria and fungi to an array of antibiotics with these convenient kits, or test your own chemical using our blank discs.
  • Convenient
  • Reliable
  • Demonstrate principles of antibiotic resistance and susceptibility
Each magazine contains 50 discs impregnated with antibiotic; all you need to do is place the disc on your culture, incubate and observe the results to determine susceptibility or resistance to specific antibiotics.
A disc dispenser, required, is available separately. Dispenser for Sensi-Disc™ releases one antibiotic disc at a time from a Sensi-Disc™ magazine.
Gram Negative Kit comes with one magazine of each of Chloramphenicol, Kanamycin, Tetracycline, Sulfamethoxazole, Nalidixic Acid, and Nitrofurantoin
Gram Positive Kit comes with one magazine of each of Penicillin, Erythromycin, Chloramphenicol, Novobiocin, Tetracycline, and Streptomycin.
Ordering information: Refrigerate upon arrival.