Ward's® Live Geobacillus stearothermophilus Culture
Supplier: Avantor
470179-120EA 13.95 USD
470179-120 470179-484
Ward's® Live Geobacillus stearothermophilus Culture
Specimens Live Specimens Bacteria - Living Material
This spore-forming, thermophilic organism is commonly used for germicidal and sporicidal testing due to its ability to withstand higher-than-normal temperatures.
  • Biosafety Level: 1
  • Gram Stain: Positive
  • Cell Morphology: Rods
  • Motility: Positive
  • Respiration: Aerobic
  • Temperature: Thermophilic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage. Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: While this organism is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.