Friction Box

Supplier: TONAWANDA MANUFACTURING
6987200
470201-438EA 39.35 USD
470201-438
Friction Box
Educational Materials Physics Educational Materials Mechanics Learning Activities
This friction box features a classic design and sturdy construction.

  • Classic design
  • Sturdy construction
  • Length: 15 cm
  • Width: 9 cm
  • Height: 8 cm

A classic wooden box for determining the coefficient of friction, the box has a ring at one end.

It measures 8×9×15 cm.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR