Ward's® Live Fiddler Crabs (Uca sp.)
Supplier: PHILIP BRYAN WOODARD
877574 877575
470180-530PK 29.99 USD
470180-530 470180-532 470006-392
Ward's® Live Fiddler Crabs (Uca sp.)
Specimens Live Specimens Aquaria - Living Material
Uca sp. The fiddler crab gets its name from the male’s larger claw, which it waves up and down in a fiddling motion when threatened or when attracting a mate.
  • Excellent for observations of circadian rhythm, regeneration and mating behavior.
  • Excellent classroom pet
Commonly found along beaches and mud flats, they act as detritivores, eating decaying matter. Crabs are approximately 1" in size.
Habitat includes:
A plastic aquarium (16"L x 9"W x 10.5"H)
Food and dish, polystyrene tray, sea salts
sand, and a coupon redeemable for a 3-pack of hermit crabs.
More Product Information