Ward's® Eye Anatomy Museum Mount
Supplier: VWR
Ward's®
WARD470000-340
470000-340EA 883.59 USD
470000-340
Ward's® Eye Anatomy Museum Mount
Specimens Preserved Specimens Display Specimens
Museum mounts are the ideal supplement for classroom dissection.
  • High quality display mount
  • Dissection reference for students
  • Quiz mechanism for teachers
Several serial sections reveal remarkable details.
Museum Mounts are expertly crafted by our own Laboratory Technicians to show external and internal anatomy in great detail. Specimens are encased in acrylic with formalin for indefinite preservation.
Museum mounts can be used as a reference for teachers and students or as a quiz mechanism by covering up the key on the back of the case.
Key includes: 1. Optic Nerve; 2. Sclera; 3. Cornea; 4. Pupil; 5. Lens; 6. Iris; 7. Retina; 8. Choroid.