Ethylene Glycol

Supplier: ALDON CORP SE

EE0110-3.8L EE0110-500ML
470301-082EA 127.45 USD
470301-082 470301-084
Ethylene Glycol
Chemicals for Science Education

CAS Number: 107-21-1
Formula Weight: 62.07
Formula: HOCH2CH2OH
Hazard Info: Toxic
Density (g/mL): 1.1135
Boiling Point (°C): 198
Freezing Point (°C): -13
Solubility: Water, Alcohol and Acetone
Synonyms: 1,2-Ethanediol
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR