Eppendorf® Pipettes
Supplier: Eppendorf
3123000020 3123000047 3123000063
470202-592EA 675.9 USD
470202-592 470202-594 470202-596
Eppendorf® Pipettes
Pipettes Micropipettes
Precise results every time.
  • Digital display
  • Adjustable volume
  • Simple calibration and maintenance
  • Easy tip ejection
Eppendorf digital micropipettes are the most convenient instruments available for pipetting small amounts of liquid. Pipettes are trim, lightweight, and specially designed not to roll on flat surfaces; the lower portions are autoclavable. Volumes can be adjusted in increments of 0.1 or 1.0 μl.