Ward's® Live Enterobacter cloacae Culture
Supplier: Avantor
470179-118EA 13.95 USD
470179-118 470179-506
Ward's® Live Enterobacter cloacae Culture
Specimens Live Specimens Bacteria - Living Material
This organism is commonly isolated from sewage, water, soil and dairy products, as it is a normal part of intestinal flora.
  • Biosafety Level: 1
  • Gram Stain: Negative
  • Cell Morphology: Rods
  • Motility: Positive
  • Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage. Freeze-dried organisms come with everything you need toMore Product Information