Ward's® Live Elodea canadensis Aquatic Plants
Supplier: EVERWOOD CO.
BUNCH
470177-082PK 17.95 USD
470177-082 470180-088 470180-090 470180-092 470180-094
Ward's® Live Elodea canadensis Aquatic Plants
Specimens Live Specimens Plants - Living Material
An excellent aquarium oxygenator, Elodea canadensis is great for teaching photosynthesis and plasmolysis.
  • Sprigs or tips are useful to demonstrate photosynthesis or plasmolysis
  • Float in your aquarium as an oxygenating hiding place for small animals
Excellent alternative if Elodea densa or Egeria najas are restricted in your state. This aquatic monocot is slightly smaller than Elodea densa and does not grow as straight sprigs, but curls into bundles.
Tips are not recommended for aquarium use. Instead, sprigs may be floated in aquaria as a food source or hiding place for fish and snails.
Ordering information: May be restricted in some states, check with your state's Department of Agriculture for restrictions. Elodea densa or Egeria najas are also available as alternatives.
Delivery information: These specimens are perishable and will ship via 2-day delivery. Please provide a requested delivery date.
Packaging: Plants are provided in waterproof, plastic bags that keep plants both fresh and healthy. Care instructions available online.