ElectroCorr LED Conductivity Indicator with Flexible Leads

Supplier: ELECTROCORR
9400500
470013-590EA 36.6 USD
470013-590
ElectroCorr LED Conductivity Indicator with Flexible Leads
Educational Materials Chemistry Educational Materials Electrochemistry Learning Activities
The ElectroCorr LED Conductivity Indicator with Flexible Leads includes a multitude of user-friendly features.

  • Length: 1¼"
  • Width: 3/4"
  • Height: 3"
  • Durable Plastic Case Protects Devices
  • Double-Insulated Wire Joint Connection

The device is protected from corrosion by a durable plastic case. Its flexible leads feature a double-insulated wire joint connection, and the unit’s LED illumination becomes more intense with increased conductivity.

The indicator is 3/4 x 1¼ x 3", and it requires a 9V battery, not included.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR