Crystal Violet

Supplier: ALDON CORP SE

CC0456-500ML CC0455-100ML CC0461-100ML CC0461-500ML CC0450-10G CC0450-25G
470300-938EA 58.1 USD
470300-938 470300-940 470300-942 470300-944 470300-946 470300-948
Crystal Violet
Chemicals for Science Education

CAS Number: 548-62-9
Formula: C25H30CIN3
Freezing Point: Decomposes
Solubility: Alcohol and Water
Synonyms: Gentian Violet, Basic Violet 3, C.I. No. 42555
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is flammable and toxic.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR