Ward's® Live Corynebacterium pseudodiphtheriticum Culture
Supplier: Avantor
470179-114EA 13.95 USD
470179-114 470179-500
Ward's® Live Corynebacterium pseudodiphtheriticum Culture
Specimens Live Specimens Bacteria - Living Material
This organism is commonly isolated from human nasopharyngeal mucosa
  • Biosafety Level: 1
  • Canada: Containment Level 2 - PHAC Licence required
  • Gram Stain: Positive
  • Cell Morphology: Irregular rods
  • Motility: Negative
  • Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage. Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: While this organism is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.