Corallina
Supplier: VWR
863938 WARD470179-982
470179-984PK 99.95 USD
470179-984 470179-982
Corallina
Specimens Live Specimens Protists - Living Material
Marine. Macroalga; pink, coral-like appearance. Produces internal frame from calcium deposits. Collected.
Characterized by reddish plastids (rhodoplasts).