Ward's® Comprehensive Animal Tissue Culture Kit
Supplier: Avantor
470024-476KT 325.19 USD
470024-476
Ward's® Comprehensive Animal Tissue Culture Kit
Cell Culture Media
All Three Culture Kits in One Package
Initiating primary cell culture from fresh embryonic tissue and observing the growth, fixing and staining cells, and comparing cell and organ cultures can all be accomplished with one complete kit. These activities, ideal for advanced high school and college classes, can be completed in three one-hour lab periods. Note: Coupon included for perishable materials. Redeem by mail, fax, phone, or e-mail.