Ward's® Metamorphosis Museum Mount
Supplier: VWR
Ward's®
WARD470000-288
470000-288EA 454.99 USD
470000-288
Ward's® Metamorphosis Museum Mount
Specimens Preserved Specimens Display Specimens
Museum mounts are the ideal supplement for classroom dissection.
  • High quality display mount
  • Dissection reference for students
  • Quiz mechanism for teachers
Perfect for insect life cycle studies.
Museum Mounts are expertly crafted by our own Laboratory Technicians to show external and internal anatomy in great detail. Specimens are encased in acrylic with formalin for indefinite preservation.
Museum mounts can be used as a reference for teachers and students or as a quiz mechanism by covering up the key on the back of the case.