Ward's® Live Chaos (Pelomyxa sp.) Culture
Supplier: Avantor
470176-448EA 9.95 USD
470176-448 470176-574 470176-576 470180-136
Ward's® Live Chaos (Pelomyxa sp.) Culture
Specimens Live Specimens Protists - Living Material
Members of the Pelomyxa genus look very much like Amoeba. Pelomyxa, however, have many nuclei, whereas Amoeba have only one.
Vital-Staining is available for this culture; cytoplasmic elements are stained blue on your live culture, to allow for better visibility.
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery information: Please specify a delivery date during checkout.