Calcium Oxide

Supplier: ALDON CORP SE

CC0120-100G CC0120-500G CC0120-500GR
470045-608EA 9.05 USD
470045-608 470300-626 470300-628 470300-630
Calcium Oxide
Chemicals for Science Education

CAS Number: 1305-78-8
Formula Weight: 56.08
Formula: CaO
Hazard Info: Irritant (produces heat with water)
Density (g/mL): 3.2-3.4
Boiling Point (°C): 2850
Freezing Point (°C): 2572
Solubility: Acids and reacts with Water
Synonyms: Lime, Quick Lime, Burnt Lime
Shelf Life (months): 12
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR