Boston Fern
Supplier: CHASE GREENHOUSES, INC.
865550
470176-222EA 12.3 USD
470176-222
Boston Fern
Specimens Live Specimens Plants - Living Material
Nephrolepis bostoniensis. Often used for plant tissue culture experiments and for demonstrating the structure and growth habits of a typical fern. It is widely grown in North America. It is supplied in a pot with care instructions. Size: approximately 6".