Ammonium Oxalate Monohydrate

Supplier: ALDON CORP SE

AA0305-100G AA0305-500G AA0305-100GR AA0304-500ML
470300-238EA 19.8 USD
470300-238 470300-240 470300-242 470225-768
Ammonium Oxalate Monohydrate
Chemicals for Science Education

CAS Number: 6009-70-7
Formula Weight: 142.11
Formula: (NH4)2C2O4·H2O
Hazard Info: Toxic
Density (g/mL): 1.5
Freezing Point (°C): 70
Solubility: Water
Synonyms: Ethanedioic Acid Diammonium Salt
Shelf Life (months): 36
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR