Ammonia

Supplier: ALDON CORP SE

AA0210-500ML AA0210-1L AA0208-500ML
470045-568EA 9.4 USD
470045-568 470300-172 470225-726
Ammonia
Chemicals for Science Education

CAS Number: Mixture
Formula Weight: Mixture
Formula: Mixture
Density: Approximately 1.0 g/mL
Boiling Point: Approximately 100°C
Freezing Point: Approximately 0°C
Solubility: Water
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: Irritant and corrosive.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR