Albumin (Egg)
Supplier: ALDON CORP SE
AA0095-100G
AA0095-500G
470300-102EA
26.1
USD
470300-102
470300-104
Albumin (Egg)
Chemicals for Science Education
CAS Number: 9006-59-1
Formula: Natural Product
Density: 1.035 g/mL
Boiling and Freezing Point: Decomposes
Solubility: Insoluble in Water
Shelf Life: 12 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...