African Violet
Supplier: CHASE GREENHOUSES, INC.
866810
470180-014EA 12.35 USD
470180-014
African Violet
Specimens Live Specimens Plants - Living Material
Saintpaulia ionatha. Dicot. This attractive, popular house plant adapts easily to a wide variety of environmental conditions, making it ideal for the classroom or a terrarium. It can be used for vegetative propagation and tissue culture and is supplied in a pot with care instructions. Size: 3–5" dia.