Acetic Anhydride
Supplier: ALDON CORP SE
CAS Number: 108-24-7
Formula Weight: 102.09
Formula: C4H6O3
Hazard Info: Irritant, Corrosive, Flammable
Density (g/mL): 1.08
Boiling Point (°C): 139.9
Freezing Point (°C): -73
Solubility: Reacts with Water and Alcohol
Synonyms: Acetic Oxide, Acetyl Oxide, Ethanoic Anhydride, Acetic Acid Anhydride
Shelf Life (months): 36
Storage: White
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...