Boreal2 Stereomicroscope - EM Series
Supplier: VWR
1X SKS LED SKS-20
470014-656EA 320.79 USD
470014-656 470039-602
Boreal2 Stereomicroscope - EM Series
Microscopes
Ideal for younger students
  • 2X objectives
  • Optional 1X, 3X, and 4X objectives available
  • Boreal Lifetime Warranty
This stereomicroscope offers a long range of focus for large and small samples. An 80 mm working distance is offered at 20X magnification for versatility in specimen studies. To protect the microscope, black, white, and translucent stage plates fit on the sealed stage to keep liquids out of the base. The ergonomic design includes a carry handle for easy transport and a large metal body with stable base for sturdy durability.