L-Threonine ≥98.5%, crystals FCC

Supplier: Spectrum Chemical Mfg. Corp.

T1057-100GM T1057-25GM T1057-1KG
95034-078EA 274.05 USD
95034-078 95034-082 95034-080
L-Threonine ≥98.5%, crystals FCC
L-Threonine

L-Threonine, Crystal, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₄H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD00064270
CAS Number: 72-19-5
EINECS: 200-774-1

Order Now

Specification Test Results

Assay 98.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR