Terpineol, mixture of isomers, Practical grade suitable for use as excipient
Supplier: SPECTRUM CHEMICAL MFG CORP
T1039-20LTBL
T1039-500MLG
T1039-100ML
T1039-4LTGL
700003-106EA
1414.35
USD
700003-106
700003-110
700003-104
700003-108
Terpineol, mixture of isomers, Practical grade suitable for use as excipient
Terpineol, mixture of isomers
Terpineol, Practical, also known as Terpene alcohol and alpha-Terpineol, is a natural monoterpene alcohol that is isolated from several sources like pine oil, cajuput oil, and petitgrain oil. Terpineol has a pleasant odor similar to lilac and is a common ingredient in perfumes, cosmetics, and flavors.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₀H₁₈O Storage Temperature: Ambient |
MDL Number:
MFCD00166983 CAS Number: 8000-41-7 EINECS: 232-268-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...