Sodium mercaptoacetate (sodium thioglycolate) ≥98%, powder
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Thioglycolic Acid
, Mercaptoacetic acid, sodium salt
, Thioglycolic Acid Sodium Salt
SO209-25GM
SO209-100GM
SO209-10KGBL
SO209-500GM
SO209-2.5KG
700008-186EA
142.03
USD
700008-186
700008-184
700008-188
700008-182
700008-180
Sodium mercaptoacetate (sodium thioglycolate) ≥98%, powder
Sodium mercaptoacetate (Sodium thioglycolate)
Sodium Thioglycolate, Powder is an organic compound and is also known as mercaptoacetic acid. It is colorless and contains thiol and carboxylic acid and has an unpleasant scent.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
HSCH₂COONa MW: 114.1 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00043386 CAS Number: 367-51-1 EINECS: 206-696-4 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...