Potassium chlorate ≥99%, crystals ACS

Supplier: Spectrum Chemical Mfg. Corp.
Danger

P1245-125GM P1245-50KGBL P1245-12KG P1245-2.5KG P1245-500GM P1245-25GM
700001-356EA 197.45 USD
700001-356 700001-366 700001-358 700001-360 700001-364 700001-362
Potassium chlorate ≥99%, crystals ACS
Potassium chlorate

Potassium Chlorate, Crystal, Reagent, ACS has been used as an oxidizing agent, a disinfectant, in oxygen preparation, and in the detection of aluminum. This simple ionic salt can yield free chlorate ions in solution which is thought to be the species responsible for this versatile activity. As an ACS grade Reagent, Spectrum Chemical manufactured compounds are used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness

Formula: KClO₃
Boiling Pt: 400 °C (1013 hPa)
Melting Pt: 356 °C
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011361
CAS Number: 3811-04-9
EINECS: 223-289-7
UN: 1485
ADR: 5.1,II
Merck Index: 13,07703

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR