Mycophenolic acid ≥98% for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: (E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoic acid

M3319-250MG M3319-1GM
700008-310EA 381 USD
700008-310 700008-308
Mycophenolic acid ≥98% for synthesis
Mycophenolic acid

Mycophenolic Acid, an immunosuppressant drug, is commonly used to prevent rejection in organ transplantation.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₇H₂₀O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00036814
CAS Number: 24280-93-1
EINECS: 246-119-3

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR