Denatonium benzoate, anhydrous 99,5, granular NF suitable for use as excipient
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
N-Benzyl-2-((2,6-dimethylphenyl)amino)-N,N-diethyl-2-oxoethanaminium benzoate
DE101-1GM
DE101-1KGBL
DE101-5KGBL
DE101-25GM
DE101-5GM
95032-806EA
92.46
USD
95032-806
95032-808
89078-784
95032-810
95032-812
Denatonium benzoate, anhydrous 99,5, granular NF suitable for use as excipient
Denatonium benzoate
Denatonium Benzoate, Anhydrous, Granular, NF is used as a denaturant and masking agent.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formula:
C₂₈H₃₄N₂O₃ MW: 446.59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00031578 CAS Number: 3734-33-6 EINECS: 223-095-2 |
Specification Test Results
Assay | 99.5% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...