Calcium bis(dihydrogenorthophosphate) monohydrate, powder FCC suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Calcium phosphate, monobasic, monohydrate

CA158-500GM CA158-125GM CA158-12KG CA158-2.5KG CA158-45KGBL
700001-730EA 164.94 USD
700001-730 700001-722 700001-724 700001-726 700001-728
Calcium bis(dihydrogenorthophosphate) monohydrate, powder FCC suitable for use as excipient
Calcium bis(dihydrogenorthophosphate) monohydrate

Calcium Phosphate, Monobasic, Monohydrate, Powder, FCC is used in the food industry as a leavening agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: H₄CaO₈P₂·1H₂O
MW: 252.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149602
CAS Number: 10031-30-8
EINECS: 231-837-1

Order Now

Specification Test Results

Assay 15.9% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR