Brij® S20

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Polyoxyethylene (20) stearyl etherSteareth 20Polyethylene glycol octadecyl ether

BR107-12KG BR107-25GM BR107-2.5KG BR107-500GM
700010-934EA 106 USD
700010-934 700010-932 700010-930 700010-938
Brij® S20
Brij® 35

Brij(R) S20 is a docosaethylene glycol mono octadecyl ether and finds use as an absorption enhancer of nonapeptide drugs

Formula: C₁₈H₃₇(OCH₂CH₂)₂₀OH
Storage Temperature: Ambient
MDL Number: MFCD00074177
CAS Number: 9005-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR