Adapter, Straight, Ace Glass Incorporated
Supplier: Ace Glass
7805-12
89060-454EA 116.96 USD
89060-454
Adapter, Straight, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features two standard taper outer joints, one with a drip tube.
  • Top and bottom [ST] outer joints
  • Drip tube
  • Fabricated from borosilicate glass
Adapter has two standard taper 14/20 outer joints, one with a 7mm drip tube.