Adapter, Connecting, Ace Glass Incorporated
Supplier: Ace Glass
5127-04 5127-06 5127-20
89082-514EA 78.13 USD
89082-514 89082-520 89082-524
Adapter, Connecting, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter is used with vacuum manifolds as port connections.
  • O-ring joint on one end
  • Outer [ST] joint on opposite end
  • One o-ring included
  • Fabricated from borosilicate glass
Featuring a #15 o-ring joint on one end and an outer standard taper joint on the opposite end. Supplied with one o-ring.