Bushing, Nylon or PTFE, Ace Glass Incorporated
Supplier: Ace Glass
5029-05 5029-10 5029-12 5029-20 5029-200 5029-202 5029-30 5029-31 5029-35 5029-40 5029-45 5029-48
89051-120EA 5.46 USD
89051-120 89051-122 89051-124 89051-126 89051-128 89051-130 89051-132 89051-134 89051-136 89051-138 89051-140 89051-142
Bushing, Nylon or PTFE, Ace Glass Incorporated
Fittings and Connectors
This is a machine threaded nylon or PTFE bushing
Available with a hole in center or solid. For use with apparatus with a #7 internal threads.
Ordering information: Solid bushing is for drilling different size hole, only. NOT intended for use as a stopper. Nylon version has a maximum temperature of 100°C. PTFE version has a maximum temperature of 160°C.
Order Now
SPECIFICATIONS
Environmentally Preferable