Precoiled Pipe Markers 'Hydrogen', Black/Yellow, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
A4064 B4064 C4064 D4064 E4064 F4064 G4064 H4064
94003-408EA 17.79 USD
94003-408 94004-182 94005-612 94009-546 94012-670 94013-866 94014-412 94015-192
Precoiled Pipe Markers 'Hydrogen', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007